Skip to Content
Merck
All Photos(1)

Key Documents

1478083

USP

R-Ofloxacin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(3R)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid, D-Levofloxacin, D-Ofloxacin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€1,230.00

€1,230.00


Estimated to ship on04 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€1,230.00

About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€1,230.00


Estimated to ship on04 April 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

ofloxacin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Fc1c(c3c4c(c1)C(=O)C(=CN4[C@@H](CO3)C)C(=O)[O-])N2CC[N+H](CC2)C

InChI

1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)/t10-/m1/s1

InChI key

GSDSWSVVBLHKDQ-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

R-Ofloxacin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service