Skip to Content
Merck
All Photos(1)

Key Documents

1357056

USP

Leflunomide Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Cyano-3-hydroxy-N-(4-trifluoromethylphenyl)crotonamide

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
€1,330.00

€1,330.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
€1,330.00

About This Item

Empirical Formula (Hill Notation):
C12H9F3N2O2
CAS Number:
Molecular Weight:
270.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€1,330.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

leflunomide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C(/C(C#N)=C(O)/C)NC1=CC=C(C(F)(F)F)C=C1

InChI

1S/C12H9F3N2O2/c1-7(18)10(6-16)11(19)17-9-4-2-8(3-5-9)12(13,14)15/h2-5,18H,1H3,(H,17,19)/b10-7-

InChI key

UTNUDOFZCWSZMS-YFHOEESVSA-N

Gene Information

human ... DHODH(1723)

Looking for similar products? Visit Product Comparison Guide

General description

Leflunomide Related Compound B (2-Cyano-3-hydroxy-N-(4-trifluoromethylphenyl)crotonamide is a malononitrilamide which is a water soluble metabolite of Leflunomide. It is considered as a noncompetitive inhibitor of dihydro-orotate dehydrogensae. This metabolite has been reported to initially target activated lymphocytes.[1]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeo Sato et al.
Rheumatology (Oxford, England), 48(10), 1265-1268 (2009-08-05)
To elucidate the factors associated with poor prognosis of LEF-induced lung injury in patients with RA. The background and clinical and laboratory features of LEF-induced lung injury were examined and compared between patients who died of and who recovered from
Yu-cong Yuan et al.
Zhonghua yi xue za zhi, 91(10), 703-706 (2011-05-24)
To explore the therapeutic effects of leflunomide metabolite A771726 on high glucose-induced podocyte injury and understand its mechanism. The conditionally immortal human glomerular podocytes were divided into normal glucose group (NG), high glucose group (HG), mannitol group (MA), high glucose
Li-Xia Xiong et al.
Xi bao yu fen zi mian yi xue za zhi = Chinese journal of cellular and molecular immunology, 25(1), 16-19 (2009-01-08)
To observe the inhibitory effects of tyrosine kinases inhibitor A77 1726 on collagen generation induced by IL-13 in fibroblasts. The inhibition rate of fibroblast proliferation with different concentration of A77 1726 was observed by MTT method. The fibroblasts were divided
Paola Montagna et al.
Annals of the New York Academy of Sciences, 1193, 30-35 (2010-04-20)
Rheumatoid arthritis (RA) prevalence is greater in females than in males, supporting estrogens as modulators of immune response. Leflunomide (LEF) is employed in the RA treatment. We studied the combinatory effects of LEF active metabolite A77 1726 (LEF-M) and 17beta-estradiol
Titia E Vrenken et al.
Journal of hepatology, 49(5), 799-809 (2008-09-24)
Leflunomide is used in the treatment of autoimmune diseases as an anti-inflammatory agent. Leflunomide and its active metabolite A77 1726 modulate mitogen-activated protein kinases (MAPK), Src kinases, the phosphoinositide-3 kinase (PI3K)/Akt-pathway and nuclear factor (NF)-kappaB activation. Both cell protective and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service