Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
500 mg | Check Cart for Availability | € 399,00 |
About This Item
Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1099477
grade
pharmaceutical primary standard
agency
USP
API family
lauric acid
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CCCCCCCCCCCC(O)=O
InChI
1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)
InChI key
POULHZVOKOAJMA-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Lauric acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Myristic Acid
- Sodium Lauroyl Sarcosinate
Biochem/physiol Actions
Substrate for CYP 4A11
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename - |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade - | grade - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) - | application(s) - |
| format neat | format neat | format - | format - |
| API family lauric acid | API family lauric acid | API family - | API family - |
Still not finding the right product?
Explore all of our products under Lauric acid
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
348.8 °F - closed cup
flash_point_c
176 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


