Skip to Content
Merck

Skip To

1354900

USP

Kaempferol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,4′,5,7-Tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Robigenin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
15 mg
Check Cart for Availability
€ 399,00

About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
304401

€ 399,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

kaempferol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C2=C(O)C(=O)c3c(O)cc(O)cc3O2

InChI

1S/C15H10O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,16-18,20H

InChI key

IYRMWMYZSQPJKC-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chromogenic reagent for antimony in the low ppm range and for gallium and indium in the sub-ppm range.
Kaempferol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ginkgo
  • Ginkgo Capsules
  • Ginkgo Tablets
  • Powdered Ginkgo Extract
  • Quercetin

Biochem/physiol Actions

Potent inhibitor of osteoclastic bone resorption. The effect is believed to be attributable to both the antioxidant and estrogenic activities of kaempferol.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2705PHL8923596353
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

primary reference standard

grade

analytical standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

PhytoLab

manufacturer/tradename

-

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

-

application(s)

food and beverages

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

API family

kaempferol

API family

kaempferol

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Kaempferol


signalword

Warning

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardExclamation mark

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters