Skip to Content
Merck

Skip To

1348532

USP

Isoeugenol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Methoxy-4-propenylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.2 mL
Check Cart for Availability
€ 469,00

About This Item

Linear Formula:
CH3OC6H3(CH=CHCH3)OH
CAS Number:
Molecular Weight:
164.20
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1909602
MDL number:

€ 469,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

API family

isoeugenol

packaging

pkg of 0.2 mL

manufacturer/tradename

USP

refractive index

n20/D 1.575 (lit.)

bp

132 °C/10 mmHg (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

OC1=CC=C(/C=C/C)C=C1OC

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3-7,11H,1-2H3/b4-3+

InChI key

BJIOGJUNALELMI-ONEGZZNKSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Isoeugenol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
34038W246808W246830
application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

application(s)

flavors and fragrances

application(s)

flavors and fragrances

format

neat

format

neat

format

-

format

-

storage temp.

−20°C

storage temp.

-

storage temp.

-

storage temp.

-

packaging

pkg of 0.2 mL

packaging

-

packaging

-

packaging

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

vapor density

>1 (vs air)

vapor density

>1 (vs air)

vapor density

>1 (vs air)

vapor density

>1 (vs air)


Still not finding the right product?

Explore all of our products under Isoeugenol


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1A

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

233.6 °F - closed cup

flash_point_c

112 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters