Skip to Content
Merck

Skip To

1342300

USP

Insulin (Pork)

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Insulin from porcine pancreas, Insulin from hog pancreas

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C256H381N65O76S6
CAS Number:
Molecular Weight:
5777.54
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

form

powder

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

InChI key

NOESYZHRGYRDHS-ZYCCASTOSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Insulin (Pork) is a biologically derived insulin obtained from porcine pancreas. It shares a high degree of homology with human insulin, differing by a single amino acid. This insulin variant has been historically used in diabetes management.

The USP biologics proteins category provides critical standards and reference materials necessary for the development and manufacturing of therapeutic proteins. These proteins, which can be naturally derived or genetically engineered, are essential in treating various diseases and conditions, ensuring that they meet quality, safety, and efficacy standards.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Insulin (Pork) USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Insulin
  • Insulin Injection
  • Isophane Insulin Suspension
  • Extended Insulin Zinc Suspension
  • Prompt Insulin Zinc Suspension
  • Insulin Zinc Suspension

Biochem/physiol Actions

Regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.
Two-chain polypeptide hormone produced by the β-cells of pancreatic islets. Its molecular weight is ~5800 Da. The α and β chains are joined by two interchain disulfide bonds. The α chain contains an intrachain disulfide bond. Insulin regulates the cellular uptake, utilization, and storage of glucose, amino acids, and fatty acids and inhibits the breakdown of glycogen, protein, and fat.

Other Notes

InChI: 1S/C256H381N65O76S6/c1-29-131(23)205(313-193(337)104-258)251(391)317-204(130(21)22)247(387)288-159(75-82-200(347)348)217(357)282-156(71-78-189(262)333)221(361)308-183-116-401-402-117-184-242(382)305-178(111-323)239(379)294-162(88-123(7)8)224(364)295-168(95-140-53-61-146(327)62-54-140)227(367)283-154(69-76-187(260)331)218(358)290-161(87-122(5)6)222(362)285-158(74-81-199(345)346)220(360)302-174(101-190(263)334)234(374)298-170(97-142-57-65-148(329)66-58-142)230(370)309-182(241(381)304-176(255(396)397)103-192(265)336)115-400-399-114-181(213(353)272-107-194(338)278-153(72-79-197(341)342)216(356)281-151(51-42-84-270-256(266)267)211(351)271-108-195(339)279-166(93-138-46-36-32-37-47-138)226(366)297-167(94-139-48-38-33-39-49-139)229(369)299-171(98-143-59-67-149(330)68-60-143)237(377)320-208(136(28)326)253(393)321-85-43-52-186(321)245(385)286-152(50-40-41-83-257)214(354)277-134(26)254(394)395)311-249(389)203(129(19)20)316-235(375)164(90-125(11)12)292-228(368)169(96-141-55-63-147(328)64-56-141)296-223(363)160(86-121(3)4)289-209(349)133(25)276-215(355)157(73-80-198(343)344)287-246(386)202(128(17)18)315-236(376)165(91-126(13)14)293-232(372)173(100-145-106-269-120-275-145)301-238(378)177(110-322)280-196(340)109-273-212(352)180(113-398-403-118-185(310-243(183)383)244(384)319-207(135(27)325)252(392)306-179(112-324)240(380)318-206(132(24)30-2)250(390)312-184)307-225(365)163(89-124(9)10)291-231(371)172(99-144-105-268-119-274-144)300-219(359)155(70-77-188(261)332)284-233(373)175(102-191(264)335)303-248(388)201(127(15)16)314-210(350)150(259)92-137-44-34-31-35-45-137/h31-39,44-49,53-68,105-106,119-136,150-186,201-208,322-330H,29-30,40-43,50-52,69-104,107-118,257-259H2,1-28H3,(H2,260,331)(H2,261,332)(H2,262,333)(H2,263,334)(H2,264,335)(H2,265,336)(H,268,274)(H,269,275)(H,271,351)(H,272,353)(H,273,352)(H,276,355)(H,277,354)(H,278,338)(H,279,339)(H,280,340)(H,281,356)(H,282,357)(H,283,367)(H,284,373)(H,285,362)(H,286,385)(H,287,386)(H,288,387)(H,289,349)(H,290,358)(H,291,371)(H,292,368)(H,293,372)(H,294,379)(H,295,364)(H,296,363)(H,297,366)(H,298,374)(H,299,369)(H,300,359)(H,301,378)(H,302,360)(H,303,388)(H,304,381)(H,305,382)(H,306,392)(H,307,365)(H,308,361)(H,309,370)(H,310,383)(H,311,389)(H,312,390)(H,313,337)(H,314,350)(H,315,376)(H,316,375)(H,317,391)(H,318,380)(H,319,384)(H,320,377)(H,341,342)(H,343,344)(H,345,346)(H,347,348)(H,394,395)(H,396,397)(H4,266,267,270)/t131-,132-,133-,134-,135+,136+,150-,151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163-,164-,165-,166-,167-,168-,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-,181-,182-,183-,184-,185-,186-,201-,202-,203-,204-,205-,206-,207-,208-/m0/s1
All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Sales restrictions may apply.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y000171713421501494079
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

form

powder

form

-

form

solid

form

solid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters