Skip to Content
Merck
All Photos(1)

Key Documents

1311000

USP

Homatropine methylbromide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Homatropine methobromide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€367.00

€367.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€367.00

About This Item

Empirical Formula (Hill Notation):
C17H24BrNO3
CAS Number:
Molecular Weight:
370.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€367.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

homatropine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Br-].[N+]1(C2CCC1CC(C2)OC(=O)C(O)c3ccccc3)(C)C

InChI

1S/C17H24NO3.BrH/c1-18(2)13-8-9-14(18)11-15(10-13)21-17(20)16(19)12-6-4-3-5-7-12;/h3-7,13-16,19H,8-11H2,1-2H3;1H/q+1;/p-1

InChI key

FUFVKLQESJNNAN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Homatropine methylbromide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Homatropine Methylbromide Tablets
  • Hydrocodone Bitartrate and Homatropine Methylbromide Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Gustavo Dos Reis et al.
Stress (Amsterdam, Netherlands), 17(4), 362-372 (2014-06-07)
Restraint stress (RS) is an experimental model to study stress-related cardiovascular responses, characterized by sustained pressor and tachycardiac responses. We used pharmacologic and surgical procedures to investigate the role played by sympathetic nervous system (SNS) and parasympathetic nervous system (PSNS)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service