Skip to Content
Merck
All Photos(1)

Key Documents

1304107

USP

Heparin Sodium Molecular Weight Calibrant

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Heparin sodium salt solution

Sign Into View Organizational & Contract Pricing

Select a Size

10.3 MG
€1,470.00

€1,470.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10.3 MG
€1,470.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€1,470.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

heparin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[Na+].[S](=O)(=O)(NC1C(OC(C(C1O[S](=O)(=O)O)OC4OC(C(C(C4O[S](=O)(=O)O)O)O)C(=O)O)CO)OC2C(OC(C(C2O)O[S](=O)(=O)O)OC3C(OC(C(C3O)NC(=O)C)O)CO[S](=O)(=O)O)C(=O)O)O

InChI

1S/C26H42N2O37S5.Na/c1-4(30)27-7-9(31)13(6(56-23(7)39)3-55-67(43,44)45)58-26-19(65-70(52,53)54)12(34)16(20(62-26)22(37)38)60-24-8(28-66(40,41)42)15(63-68(46,47)48)14(5(2-29)57-24)59-25-18(64-69(49,50)51)11(33)10(32)17(61-25)21(35)36;/h5-20,23-26,28-29,31-

InChI key

JRTRSJGZMRQDHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heparin Sodium is a mixture of sulfated glycosaminoglycans in the form of sodium salt. It is composed of heterogeneous molecules with different molecular weights, collectively exhibiting multiple activities that affect various factors involved in the blood clotting process. This substance naturally occurs in mammalian tissues and is typically derived from sources such as the intestinal mucosa or other applicable tissues obtained from domestic mammals commonly consumed as food by humans.[1] Heparin Sodium Molecular Weight Calibrant USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Heparin Sodium Molecular Weight Calibrant USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Heparin Sodium

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heparin Sodium
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 41(6), 2195-2195 (2018)
Heparin Sodium
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(5) (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service