Skip to Content
Merck

Skip To

1301404

USP

Guanabenz acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Guanabenz acetate salt, 1-(2,6-Dichlorobenzylideneamino)guanidine acetate salt, WY-8678

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 269,00

About This Item

Empirical Formula (Hill Notation):
C8H8Cl2N4 · C2H4O2
CAS Number:
Molecular Weight:
291.13
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 269,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

guanabenz acetate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(O)=O.NC(=N)N\N=C\c1c(Cl)cccc1Cl

InChI

1S/C8H8Cl2N4.C2H4O2/c9-6-2-1-3-7(10)5(6)4-13-14-8(11)12;1-2(3)4/h1-4H,(H4,11,12,14);1H3,(H,3,4)/b13-4+;

InChI key

MCSPBPXATWBACD-GAYQJXMFSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Guanabenz acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Also, for use with USP monograph such as Guanabenz Acetate Tablets

Biochem/physiol Actions

Centrally acting α2 adrenoceptor agonist; I2 imidazoline binding site ligand; antihypertensive.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
129675612972051270800
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

guanabenz acetate

API family

gonadorelin

API family

goserelin

API family

flecainide

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

-

Gene Information

human ... GNRHR(2798)

Gene Information

human ... SCN5A(6331)


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters