Skip to Content
Merck

Skip To

1269458

USP

Fenoldopam mesylate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Fenoldopam methanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 724,00

About This Item

Empirical Formula (Hill Notation):
C16H16ClNO3 · CH4O3S
CAS Number:
Molecular Weight:
401.86
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 724,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

fenoldopam

manufacturer/tradename

USP

mp

271 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CS(O)(=O)=O.Oc1ccc(cc1)C2CNCCc3c(Cl)c(O)c(O)cc23

InChI

1S/C16H16ClNO3.CH4O3S/c17-15-11-5-6-18-8-13(9-1-3-10(19)4-2-9)12(11)7-14(20)16(15)21;1-5(2,3)4/h1-4,7,13,18-21H,5-6,8H2;1H3,(H,2,3,4)

InChI key

CVKUMNRCIJMVAR-UHFFFAOYSA-N

Gene Information

human ... DRD1(1812)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenoldopam mesylate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fenoldopam Mesylate Injection

Biochem/physiol Actions

Dopamine D1-receptor agonist; Antihypertensive
Fenoldopam mesylate is a selective dopamine D1 receptor agonist used as an antihypertensive agent. It is an agonist for D1-like dopamine receptors and binds with moderate affinity to α2-adrenoceptors. It has no significant affinity for D2-like receptors, α1 and α-adrenoceptors, 5HT1 and 5HT2 receptors, or muscarinic receptors. Fenoldopam mesylate causes peripheral vasodilation by stimulating dopamine-1receptors and α2-adrenoceptors, increasing renal blood flow.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML019812694701269469
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

neat

format

neat

mp

271 °C (lit.)

mp

271 °C (lit.)

mp

-

mp

-

Gene Information

human ... DRD1(1812)

Gene Information

human ... DRD1(1812)

Gene Information

-

Gene Information

-


Still not finding the right product?

Explore all of our products under Fenoldopam mesylate


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters