Skip to Content
Merck

Skip To

1236889

USP

Epinastine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9,13b-Dihydro-1H-dibenz[cf]imidazo[1,5-a]azepine hydrochloride, WAL-801Cl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mg
Check Cart for Availability
€ 1.180,00

About This Item

Empirical Formula (Hill Notation):
C16H15N3 · HCl
CAS Number:
Molecular Weight:
285.77
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 1.180,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

epinastine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].NC1=NCC2N1c3ccccc3Cc4ccccc24

InChI

1S/C16H15N3.ClH/c17-16-18-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)19(15)16;/h1-8,15H,9-10H2,(H2,17,18);1H

InChI key

VKXSGUIOOQPGAF-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Epinastine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Epinastine hydrochloride is a non-sedating H1 histamine receptor antagonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0001139E51561218005
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

API family

epinastine

API family

epinastine

API family

-

API family

diphenhydramine

Gene Information

human ... HRH1(3269)

Gene Information

human ... HRH1(3269)

Gene Information

human ... HRH1(3269)

Gene Information

human ... HRH1(3269)


Still not finding the right product?

Explore all of our products under Epinastine hydrochloride


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters