Skip to Content
Merck

Skip To

1225485

USP

Doxazosin Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N,N′-Bis(1,4-benzodioxane-2-carbonyl)piperazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
15 mg
Check Cart for Availability
€ 1.120,00

About This Item

Empirical Formula (Hill Notation):
C22H22N2O6
CAS Number:
Molecular Weight:
410.42
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 1.120,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

doxazosin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N3(CCN(CC3)C(=O)C4Oc5c(cccc5)OC4)C(=O)C1Oc2c(cccc2)OC1

InChI

1S/C22H22N2O6/c25-21(19-13-27-15-5-1-3-7-17(15)29-19)23-9-11-24(12-10-23)22(26)20-14-28-16-6-2-4-8-18(16)30-20/h1-8,19-20H,9-14H2

InChI key

RYWJYOGVNZEBKW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1225474PHR2954PHR3308
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

API family

doxazosin

API family

doxazosin

API family

doxazosin

API family

-


Still not finding the right product?

Explore all of our products under Doxazosin Related Compound F


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters