Skip to Content
Merck

Skip To

1220517

USP

Dipyridamole Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,2′-[(4,6,8-Tri-1-piperidinylpyrimido[5,4-d]pyrimidin-2-yl)imino]bisethanol, 2,2′-{[4,6,8-Tri(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]azanediyl}diethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 1.270,00

About This Item

Empirical Formula (Hill Notation):
C25H40N8O2
CAS Number:
Molecular Weight:
484.64
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107

€ 1.270,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

dipyridamole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
122054012205591220539
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

API family

dipyridamole

API family

dipyridamole

API family

dipyridamole

API family

dipyridamole


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters