Skip to Content
Merck

Skip To

1163253

USP

Dapagliflozin propanediol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dapagliflozin propanediol monohydrate, (1S)-1,5-Anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-D-glucitol (S)-propane-1,2-diol (1:1) monohydrate, (2S,3R,4R,5S,6R)-2-(3-(4-Ethoxybenzyl)-4-chlorophenyl)-6-hydroxymethyl-tetrahydro-2H-pyran-3,4,5-triol propanediol monohydrate, BMS 512148 propanediol hydrate, BMS-512148 propanediol hydrate, Dapagliflozin (S)-propylene glycol monohydrate, Dapagliflozin propanediol hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 1.000,00

About This Item

Empirical Formula (Hill Notation):
C21H25ClO6·C3H8O2·H2O
CAS Number:
Molecular Weight:
502.98
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

€ 1.000,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

dapagliflozin

packaging

pkg of 100 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1c(cc(cc1)[C@@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)Cc2ccc(cc2)OCC.O[C@H](CO)C.O

InChI

1S/C21H25ClO6.C3H8O2.H2O/c1-2-27-15-6-3-12(4-7-15)9-14-10-13(5-8-16(14)22)21-20(26)19(25)18(24)17(11-23)28-21;1-3(5)2-4;/h3-8,10,17-21,23-26H,2,9,11H2,1H3;3-5H,2H2,1H3;1H2/t17-,18-,19+,20-,21+;3-;/m10./s1

InChI key

GOADIQFWSVMMRJ-UPGAGZFNSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Dapagliflozin propanediol USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Dapagliflozin (BMS-512148) is an orally active, potent and selective renal sodium-dependent glucose cotransporter 2 (SGLT2; SLC5A2) inhibitor (human/rat SGLT2 IC50 = 1.1/3 nM vs. human/rat SGLT1 IC50 = 1.39/0.6 μM; R-methyl-D-glucopyranoside (AMG) uptake assay with respective CHO transfectants). Dapagliflozin reduces blood glucose levels (by 55% in streptozotocin (STZ)-induced hyperglycemic rats 5 hrs post 0.1 mg/kg p.o.) by preventing the renal glucose reabsorption process and promoting glucose excretion in urine with good pharmacokinetic properties and oral availability (84% in rats and 83% in dogs, respectively, post 1 mg/kg and 6.6 mg/kg p.o.) in vivo.
Orally active, potent and selective renal sodium-dependent glucose cotransporter 2 (SGLT2; SLC5A2) inhibitor with blood sugar-lowering efficacy in vivo.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
11632641600201SML2758
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

format

neat

format

neat

format

neat

format

-

storage temp.

2-8°C

storage temp.

room temp

storage temp.

15-25°C

storage temp.

−20°C

packaging

pkg of 100 mg

packaging

pkg of 15 mg

packaging

-

packaging

-

API family

dapagliflozin

API family

dapagliflozin

API family

regorafenib

API family

-


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters