Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 mg | Check Cart for Availability | € 1.380,00 |
About This Item
Empirical Formula (Hill Notation):
C14H12N4O6
CAS Number:
Molecular Weight:
332.27
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
agency
USP
API family
dantrolene
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Dantrolene Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Dantrolene Sodium
- Dantrolene Sodium for Injection
- Dantrolene Sodium Capsules
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade certified reference material, pharmaceutical secondary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename USP |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical small molecule | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) |
| format neat | format - | format - | format neat |
| API family dantrolene | API family dantrolene | API family dantrolene | API family dantrolene |
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



