Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Check Cart for Availability | € 1.130,00 |
About This Item
Empirical Formula (Hill Notation):
C11H10N4O
CAS Number:
Molecular Weight:
214.22
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
pharmaceutical primary standard
agency
USP
API family
dalfampridine
form
solid
manufacturer/tradename
USP
application(s)
pharmaceutical
SMILES string
N(c2ccncc2)C(=O)Nc1ccncc1
InChI
1S/C11H10N4O/c16-11(14-9-1-5-12-6-2-9)15-10-3-7-13-8-4-10/h1-8H,(H2,12,13,14,15,16)
InChI key
QRRGJASIECDXQW-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Dalfampridine Related Compound C USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Dalfampridine
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP | manufacturer/tradename USP |
| form solid | form - | form - | form - |
| application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical |
| API family dalfampridine | API family - | API family dalfampridine | API family dalfampridine |
Still not finding the right product?
Explore all of our products under Dalfampridine Related Compound C
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


