Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Check Cart for Availability | € 1.380,00 |
About This Item
Linear Formula:
(ClCH2CH2)2NH·HCl
CAS Number:
Molecular Weight:
178.49
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
3550356
MDL number:
€ 1.380,00
Check Cart for Availability
grade
pharmaceutical primary standard
agency
USP
API family
cyclophosphamide
manufacturer/tradename
USP
mp
212-214 °C (lit.)
application(s)
pharmaceutical (small molecule)
SMILES string
Cl.ClCCNCCCl
InChI
1S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1H
InChI key
YMDZDFSUDFLGMX-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk
WGK 3
flash_point_f
No information available.
flash_point_c
No information available.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

