Skip to Content

1145003

USP

Codeine sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Codeine sulfate salt, 3-Methylmorphine sulfate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C36H42N2O6 · H2SO4
CAS Number:
Molecular Weight:
696.81
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

codeine

manufacturer/tradename

USP

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[S](=O)(=O)(O)O.N6([C@H]7[C@H]8[C@@]9([C@@H](Oc%10c9c(ccc%10OC)C7)[C@H](C=C8)O)CC6)C.N1([C@H]2[C@H]3[C@@]4([C@@H](Oc5c4c(ccc5OC)C2)[C@H](C=C3)O)CC1)C

InChI

1S/2C18H21NO3.H2O4S/c2*1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19;1-5(2,3)4/h2*3-6,11-13,17,20H,7-9H2,1-2H3;(H2,1,2,3,4)/t2*11-,12+,13-,17-,18-;/m00./s1

InChI key

BCXHDORHMMZBBZ-DORFAMGDSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Codeine sulfate USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Codeine Sulfate Tablets
  • Codeine Sulfate Oral Solution
  • Dihydrocodeine Bitartrate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


Hazard Classifications

Acute Tox. 4 Oral

pictograms

Exclamation mark

signalword

Warning

hcodes

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library