Skip to Content
Merck

Skip To

1056130

USP

Benzophenone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Diphenyl ketone, Diphenylmethanone, NSC 8077

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 1.380,00

About This Item

Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1238185

€ 1.380,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor pressure

1 mmHg ( 108 °C)

API family

benzophenone

form

crystalline

manufacturer/tradename

USP

bp

305 °C (lit.)

mp

47-51 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

O=C(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

General description

Benzophenone is an aromatic ketone that serves as a photoinitiator and UV stabilizer in biologic formulations. In pharmaceutical applications, it is primarily utilized to prevent degradation of active pharmaceutical ingredients (APIs) caused by exposure to UV light. Its inclusion in biologic product formulations helps ensure the stability and longevity of these products, particularly in formulations sensitive to light-induced degradation.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzophenone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Ph
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.018017366450296
grade

pharmaceutical primary standard

grade

-

grade

analytical standard

grade

analytical standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

-

application(s)

food and beverages
pharmaceutical

application(s)

food and beverages
pharmaceutical

format

neat

format

-

format

neat

format

neat

form

crystalline

form

crystals

form

-

form

-

bp

305 °C (lit.)

bp

304-306 °C/1013 hPa

bp

305 °C (lit.)

bp

305 °C (lit.)


Still not finding the right product?

Explore all of our products under Benzophenone


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

target_organs

Liver,Kidney

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

119-61-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters