Skip to Content
Merck

Skip To

1044334

USP

Atazanavir sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dimethyl (3S,8S,9S,12S)-9-benzyl-3,12,di-tert-butyl-8-hydroxy-4,11-dioxo-6-(p-2-pyridylbenzyl)-2,5,6,10,13-pentaazatetradecanedioate sulfate (1:1)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Check Cart for Availability
€ 420,00

About This Item

Empirical Formula (Hill Notation):
C38H52N6O7 · H2SO4
CAS Number:
Molecular Weight:
802.93
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€ 420,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

atazanavir

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)([O-])[O-].N(NC(=O)[C@@H](NC(=O)OC)C(C)(C)C)(C[C@H](O)[C@@H](NC(=O)[C@@H](NC(=O)OC)C(C)(C)C)Cc3ccccc3)Cc1ccc(cc1)c2ncccc2.[H+].[H+]

InChI

1S/C38H52N6O7.H2O4S/c1-37(2,3)31(41-35(48)50-7)33(46)40-29(22-25-14-10-9-11-15-25)30(45)24-44(43-34(47)32(38(4,5)6)42-36(49)51-8)23-26-17-19-27(20-18-26)28-16-12-13-21-39-28;1-5(2,3)4/h9-21,29-32,45H,22-24H2,1-8H3,(H,40,46)(H,41,48)(H,42,49)(H,43,47);(H2,1,2,3,4)/t29-,30-,31+,32+;/m0./s1

InChI key

DQSGVVGOPRWTKI-QVFAWCHISA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atazanavir sulfate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0002059PHR3002SML2308
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical

application(s)

-

API family

atazanavir

API family

atazanavir

API family

atazanavir

API family

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Atazanavir sulfate


flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters