Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
50 mg | Check Cart for Availability | € 446,00 |
About This Item
Empirical Formula (Hill Notation):
C6H10O5
CAS Number:
Molecular Weight:
162.14
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
80998
grade
pharmaceutical primary standard
agency
USP
API family
levoglucosan
manufacturer/tradename
USP
mp
182-184 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
O[C@H]1[C@H](O)[C@H]2CO[C@H](O2)[C@@H]1O
InChI
1S/C6H10O5/c7-3-2-1-10-6(11-2)5(9)4(3)8/h2-9H,1H2/t2-,3-,4+,5-,6-/m1/s1
InChI key
TWNIBLMWSKIRAT-VFUOTHLCSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- 1,6-Anhydro-D-glucose USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
Hydrogenated Polydextrose - Polydextrose
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.