Skip to Content
Merck

Skip To

1035005

USP

Anethole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

trans-Anethole, 4-Propenylanisole, trans-1-Methoxy-4-(1-propenyl)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
4 x 0.5 mL
Check Cart for Availability
€ 440,00

About This Item

Linear Formula:
CH3CH=CHC6H4OCH3
CAS Number:
Molecular Weight:
148.20
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
774229

€ 440,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

anethole

manufacturer/tradename

USP

refractive index

n20/D 1.561 (lit.)

bp

234-237 °C (lit.)

mp

20-21 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc(\C=C\C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+

InChI key

RUVINXPYWBROJD-ONEGZZNKSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Naturally occurring phenylpropene derivative that is estrogenic at lower concentrations and cytotoxic at higher concentrations to cancer cell lines. The cytotoxicity is related to the metabolism of trans-anethole to 4-hydroxy-1-propenylbenzene.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR12188.00429W208604
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

FG, Fragrance grade, Halal, Kosher

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

flavors and fragrances

format

neat

format

neat

format

-

format

-

refractive index

n20/D 1.561 (lit.)

refractive index

n20/D 1.561 (lit.)

refractive index

-

refractive index

n20/D 1.561 (lit.)

API family

anethole

API family

anethole

API family

-

API family

-


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

213.8 °F

flash_point_c

101 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters