Skip to Content
Merck
All Photos(1)

Documents

1009479

USP

2-Acetylpyridine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl 2-pyridyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
Beilstein:
107759
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

2-acetylpyridine

manufacturer/tradename

USP

refractive index

n20/D 1.521 (lit.)

bp

188-189 °C (lit.)

density

1.08 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)c1ccccn1

InChI

1S/C7H7NO/c1-6(9)7-4-2-3-5-8-7/h2-5H,1H3

InChI key

AJKVQEKCUACUMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

2-Acetylpyridine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joëlle Akl et al.
Dalton transactions (Cambridge, England : 2003), 43(33), 12721-12733 (2014-07-12)
The 4'-(2-fluorenyl)-2,2':6',2''-terpyridine (FT) ligand and its cis(Cl,Cl)- and trans(Cl,Cl)-[Ru(II)(FT)Cl2(NO)](PF6) complexes have been synthesized. Both isomers were separated by HPLC and fully characterized by (1)H and (13)C NMR. The X-ray diffraction crystal structures were solved for FT (Pna21 space group, a
Subhashree P Dash et al.
Journal of inorganic biochemistry, 144, 1-12 (2015-01-13)
Three dioxidovanadium(V) complexes [VO2L(1-3)] (1-3) [HL(1)=1-napthoyl hydrazone of 2-acetyl pyridine, HL(2)=2-furoyl hydrazone of 2-acetyl pyridine and H2L(3)=isonicotinoyl hydrazone of 2-hydroxy benzaldehyde] have been reported. All the complexes were characterized by various spectroscopy (IR, UV-visible and NMR) and the molecular structures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service