Skip to Content
Merck
All Photos(1)

Key Documents

SY3H580D8A6F

Synthonix Corporation

trans-2-(difluoromethyl)cyclopropanecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
136.1
MDL number:

Iupac Name

(1R,2R)-2-(difluoromethyl)cyclopropane-1-carboxylic acid

Inchi Code

1S/C5H6F2O2/c6-4(7)2-1-3(2)5(8)9/h2-4H,1H2,(H,8,9)/t2-,3-/m1/s1

InChI key

AQPYFAYLGMEWCD-PWNYCUMCSA-N

storage temp.

Refrigerator

Purity

90%

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

oil

Product Link

https://synthonix.com/60171.html

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://synthonix.com/60171.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library