Y0002201
Fluoxetine impurity A
CRS, European Pharmacopoeia (EP) Reference Standard
Synonym(s):
α-[2-(Methylamino)ethyl]benzyl alcohol, 3-(Methylamino)-1-phenylpropan-1-ol
About This Item
Recommended Products
API family
fluoxetine
manufacturer/tradename
EDQM
mp
59-64 °C (lit.)
storage temp.
2-8°C
SMILES string
CNCCC(O)c1ccccc1
InChI
1S/C10H15NO/c1-11-8-7-10(12)9-5-3-2-4-6-9/h2-6,10-12H,7-8H2,1H3
InChI key
XXSDCGNHLFVSET-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service