Skip to Content
Merck
All Photos(1)

Documents

SAAR2436020

Formaldehyde solution

uniLAB

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
MDL number:
UNSPSC Code:
12352501

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

product line

uniLAB

autoignition temp.

572 °F

packaging

poly bottle of 4 × 2.5 L (SAAR2436020LP)
pkg of 25 L (SAAR2436020ME)
poly bottle of 4 × 5 L (SAAR2436020LL)

density

1.09 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service