Skip to Content
Merck
All Photos(1)

Documents

PTCO005

Supelco

Pt-Co/Hazen/APHA Colour Reference Standard

ASTM D 1209 (ASTM colour 5)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is an analytical standard certified under ISO 9001 Quality system.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Pt-Co/Hazen/APHA colour reference standards can be found on the Paragon Scientific webpage.

Application

Pt-Co/Hazen/APHA (ASTM colour 5) colour reference standard is used to routinely calibrate color measuring instruments such as colorimeters.

Features and Benefits

  • Qualified under ISO 9001 Quality management system
  • Available with a certificate of analysis and safety data sheet
  • Traceable to international standards with maximum data accuracy
  • Colour values corresponding to ASTM D 1209 colour scale
  • Delivered in 500-mL pack size with tamper-evident packaging
  • 12-months shelf life

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service