Skip to Content
Merck
All Photos(1)

Key Documents

PS379

Terbutryn

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N5S
CAS Number:
Molecular Weight:
241.36
Beilstein:
611817
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. PS-379

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NC(C)(C)C)nc(SC)n1

InChI

1S/C10H19N5S/c1-6-11-7-12-8(15-10(2,3)4)14-9(13-7)16-5/h6H2,1-5H3,(H2,11,12,13,14,15)

InChI key

IROINLKCQGIITA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terbutryn is a systemic herbicide used primarily for elimination of annual broadleaf weeds. It is a triazine compound and is known to inhibit photosynthesis. This selective herbicide is also used in controlling submerged and free-floating weeds, algae in reservoirs, and fish ponds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Terbutryn may be used as a reference standard for the determination of terbutryn in water samples by solid-phase extraction followed by high-performance liquid chromatography.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of triazines and dinitroanilines in waters by high-performance liquid chromatography after solid-phase extraction.
Vitali P, et al.
Journal of Chromatography A, 660(1-2), 219-222 (1994)
Effect of Pendimethalin, Trifluralin and Terbutryn on Lolium multiflorum growing with barley during pre-emergence stage.
Alshallash KS.
Annals of Agricultural Science, 59(2), 239-242 (2014)
C Rioboo et al.
Aquatic toxicology (Amsterdam, Netherlands), 83(4), 247-253 (2007-06-08)
Few data exist on potential toxic effects that pollutants may have on zooplankton fed microalgae exposed to pesticides. For that reason, microalgal cultures were exposed to different concentrations of the triazine herbicide terbutryn, and used as exclusive food source to
Kristin Quednow et al.
Environmental science and pollution research international, 16(6), 630-640 (2009-05-23)
The present study focuses on the temporal concentration changes of four common organic pollutants in small freshwater streams of Hesse, Germany. The substances (tris(2-chloroethyl)phosphate (TCEP), the technical isomer mixture of 4-nonylphenol (NP), 2-(t-butylamino)-4-(ethylamino)-6-(methylthio)-s-triazine (terbutryn), and N,N-diethyl-m-toluamide (DEET)) are subject to
Matthias Broser et al.
The Journal of biological chemistry, 286(18), 15964-15972 (2011-03-04)
Herbicides that target photosystem II (PSII) compete with the native electron acceptor plastoquinone for binding at the Q(B) site in the D1 subunit and thus block the electron transfer from Q(A) to Q(B). Here, we present the first crystal structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service