Skip to Content
Merck
All Photos(1)

Key Documents

PS319

2,4-D isooctyl ester

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

analytical standard

packaging

ampule of 1000 mg

manufacturer/tradename

Chem Service, Inc. PS-319

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

InChI

1S/C16H22Cl2O3/c1-3-5-6-12(4-2)10-21-16(19)11-20-15-8-7-13(17)9-14(15)18/h7-9,12H,3-6,10-11H2,1-2H3

InChI key

QZSFJRIWRPJUOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Effect of 2,4-D isooctylester on sperms in tail of epididymis and HSP70 expression of testis tissue in rats].
Guo-Yuan Chen et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 22(4), 281-282 (2004-09-10)
Congyun Liu et al.
Environmental monitoring and assessment, 184(7), 4247-4251 (2011-08-05)
A simple analytical method was developed to determine the 2,4-D isooctyl ester residue in wheat and soil by gas chromatography coupled with electron capture detector. Using the method, the dissipation and residue of 2,4-D isooctyl ester in wheat field was
J Harville et al.
American Industrial Hygiene Association journal, 50(8), 438-446 (1989-08-01)
Tyvek (laminated saranax) and unsupported nitrile gloves gave protection from permeation against formulation concentrates and aqueous emulsions of Esteron 99 [isooctyl ester of (2,4-dichlorophenoxy)acetic acid] under conditions simulating extremes of field conditions. Neoprene gloves, whether unsupported or supported, lined or
Acute toxicity of four phenoxy herbicides to aquatic organisms.
H C Alexander et al.
Bulletin of environmental contamination and toxicology, 35(3), 314-321 (1985-09-01)
R P Moody et al.
Journal of toxicology and environmental health, 29(3), 237-245 (1990-01-01)
Dermal absorption of the 14C-ring-labeled phenoxy herbicides 2,4-D [(2,4-dichlorophenoxy)acetic acid], 2,4-D amine (2,4-dichlorophenoxyacetic acid dimethylamine), 2,4-D isooctyl (2,4-dichlorophenoxyacetic acid isooctyl ester), and 2,4,5-T amine (2,4,5-trichlorophenoxyacetic acid trimethylamine) was examined following topical applications of the herbicides to the back of rabbits

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service