Skip to Content
Merck
All Photos(1)

Key Documents

PS2081

Pyridate

analytical standard

Synonym(s):

O-(6-Chloro-3-phenylpyridazin-4-yl) S-octyl carbonothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23ClN2O2S
CAS Number:
Molecular Weight:
378.92
Beilstein:
759528
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 500 mg

manufacturer/tradename

Chem Service, Inc. PS-2081

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCCCCCSC(=O)Oc1cc(Cl)nnc1-c2ccccc2

InChI

1S/C19H23ClN2O2S/c1-2-3-4-5-6-10-13-25-19(23)24-16-14-17(20)21-22-18(16)15-11-8-7-9-12-15/h7-9,11-12,14H,2-6,10,13H2,1H3

InChI key

JTZCTMAVMHRNTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Lindner et al.
International journal of environmental analytical chemistry, 16(3), 205-218 (1983-01-01)
For many reasons residue analysis of plant protecting chemicals is becoming of increasing importance. The demand of modern trace analysis techniques is to detect significantly and sensitively values down to levels in the ppb range or even less. Aggravating circumstances
A Pachinger et al.
Journal of chromatography, 558(2), 369-373 (1991-10-11)
The main metabolite of the herbicide pyridate is 3-phenyl-4-hydroxy-6-chlorpyridazine (CL9673). A high-performance liquid chromatographic method with amperometric detection is described for determining CL9673 at residue levels in water samples. Sample preconcentration is performed by passage through a C18 extraction cartridge.
William L Patzoldt et al.
Pest management science, 59(10), 1134-1142 (2003-10-17)
While surveying Illinois Amaranthus tuberculatus (Moq) Sauer (tall waterhemp) half-sib populations for herbicide response variability, several were observed to segregate for resistance to atrazine. Studies were conducted on greenhouse-grown A tuberculatus plants to compare atrazine responses among populations that were
Markus Burghardt et al.
Journal of agricultural and food chemistry, 53(18), 7150-7155 (2005-09-01)
Sorption and diffusion of the herbicide pyridate and its metabolite CL9673 were measured in reconstituted cuticular waxes isolated from Chenopodium album L. and Hordeum vulgare L. (cultivar Igri) leaves. The compounds have the same basic chemical structure, except that pyridate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service