Skip to Content
Merck
All Photos(1)

Key Documents

PS2075

Azadirachtin

technical mixture, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H44O16
CAS Number:
Molecular Weight:
720.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 10 mg

manufacturer/tradename

Chem Service, Inc. PS-2075

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[H]C1([H])C2OC3([H])OC=CC3(O)C1C4(C)OC24[C@@]5(C)[C@@H](O)[C@@H]6OC[C@]7([C@H](C[C@H](OC(=O)C(C)=CC)[C@]8(COC(O)([C@H]58)C(=O)OC)[C@@]67[H])OC(C)=O)C(=O)OC

InChI

1S/C35H44O16/c1-8-15(2)24(38)49-18-12-19(48-16(3)36)32(26(39)43-6)13-46-21-22(32)31(18)14-47-34(42,27(40)44-7)25(31)29(4,23(21)37)35-20-11-17(30(35,5)51-35)33(41)9-10-45-28(33)50-20/h8-10,17-23,25,28,37,41-42H,11-14H2,1-7H3/t17?,18-,19?,20?,21+,22+,23?,25?,28?,29?,30?,31?,32-,33?,34-,35?/m0/s1

InChI key

FTNJWQUOZFUQQJ-CFYAVYSVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crude extract of the neem tree. Not intended for use as a quantitative standard

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Triterpenoid found in need tree seeds, azadiractin suppresses feeding by many insect species and disrupts growth of most insect and other arthropod species, while having very low mammalian toxicity. Promising as a natural pesticide.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Cortez et al.
Experimental parasitology, 131(3), 363-371 (2012-05-26)
The effects of azadirachtin and ecdysone on the Trypanosoma cruzi population in the Rhodnius prolixus gut were investigated. T. cruzi were rarely found in the gut compartments of azadirachtin-treated larvae. High parasite numbers were observed in the stomach of the
Pasquale Mosesso et al.
Toxicology letters, 213(3), 361-366 (2012-08-14)
In this work we have examined the genotoxic potential of the bioinsecticide Azadirachtin A (AZA) and its influence on cell proliferation on human lymphocytes and Chinese Hamster ovary (CHO) cells. AZA genotoxicity was assessed by the analysis of chromosomal aberrations
Saurav Kumar et al.
Fish & shellfish immunology, 34(2), 564-573 (2012-12-25)
Modulation of the immune responses using active bio-ingredients as a possible prophylaxis measure has been novel prospect for aquaculture. The present study evaluated the effects of azadirachtin EC 25% on non-specific immune responses in goldfish Carassius auratus and resistance against
Smita Srivastava et al.
Applied biochemistry and biotechnology, 166(2), 365-378 (2011-11-16)
Azadirachtin, a well-known biopesticide is a secondary metabolite conventionally extracted from the seeds of Azadirachta indica. The present study involved in vitro azadirachtin production by developing hairy roots of A. indica via Agrobacterium rhizogenes-mediated transformation of A. indica explants. Liquid
Smita Srivastava et al.
Bioprocess and biosystems engineering, 35(9), 1549-1553 (2012-05-15)
Present investigation involves hairy root cultivation of Azadirachta indica in a modified stirred tank reactor under optimized culture conditions for maximum volumetric productivity of azadirachtin. The selected hairy root line (Az-35) was induced via Agrobacterium rhizogenes LBA 920-mediated transformation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service