Skip to Content
Merck
All Photos(1)

Key Documents

PHL89624

Dehydrocostus lactone

phyproof® Reference Substance

Synonym(s):

(−)-Dehydrocostus lactone; Dehydrocostus lactone; Epiligulyl oxide, (3aS,6aR,9aR,9bS)-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3H)-one, Dehydro-costus lactone (6CI,7CI), Dehydrocostuslactone, Guaia-4(15),10(14),11(13)-trien-12-oic acid, 6alpha-hydroxy-, gamma-lactone, [3aS-(3aα,6aα,9aα,9bβ)]-Decahydro-3,6,9-tris(methylene)-azuleno[4,5-b]furan-2(3H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€394.00

€394.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€394.00

About This Item

Empirical Formula (Hill Notation):
C15H18O2
CAS Number:
Molecular Weight:
230.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€394.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

SMILES string

C=C1CC[C@@]2([H])C(CC[C@@H]3[C@H](OC(C3=C)=O)[C@]21[H])=C

InChI

1S/C15H18O2/c1-8-4-7-12-10(3)15(16)17-14(12)13-9(2)5-6-11(8)13/h11-14H,1-7H2/t11-,12-,13-,14-/m0/s1

InChI key

NETSQGRTUNRXEO-XUXIUFHCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Dehydrocostuslactone (DHE) is a sesquiterpene lactone extracted from Saussurea lappa and Aucklandia lappa. The compound possess antifungal and antimycobacterial activity and exhibit a cytotoxic effect against HepG2, OVCAR-3, and HeLa cell lines.
Suppressors of cytokine signaling (SOCS) proteins negatively regulate cytokine receptor signaling. The SOCS family include eight members SOCS-1 to SOCS-7 and CIS. Dehydrocostuslactone (DHE) is a sesquiterpene lactone extracted from Saussurea lappa and Aucklandia lappa. The compound possess antifungal and antimycobacterial activity and exhibit a cytotoxic effect against HepG2, OVCAR-3, and HeLa cell lines. DHE inhibits cell proliferation not only in early-stage breast cancer MCF-7 but also in more advanced stage MDA-MB-231 cancer cells, without significant effect on H184B5F5/M10 normal mammary epithelial cells. The inhibition of breast cancer cells proliferation occurs through cell cycle blockade and apoptosis. DHE inhibits cell cycle progression and cell death through SOCS-1 and SOCS-3.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service