Skip to Content
Merck
All Photos(1)

Key Documents

PHL89587

Bufalin

phyproof® Reference Substance

Synonym(s):

3β,14-Dihydroxy-5β,20(22)-bufadienolide, 5β,20(22)-Bufadienolide-3β,14-diol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€428.00

€428.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€428.00

About This Item

Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
UNSPSC Code:
41116107
NACRES:
NA.24

€428.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@]([C@@](CC[C@@]1([H])[C@@]2(CC[C@H](O)C1)C)([H])[C@]2([H])CC3)(CC[C@@H]4C(C=C5)=COC5=O)[C@]34C

InChI

1S/C24H34O4/c1-22-10-7-17(25)13-16(22)4-5-20-19(22)8-11-23(2)18(9-12-24(20,23)27)15-3-6-21(26)28-14-15/h3,6,14,16-20,25,27H,4-5,7-13H2,1-2H3/t16-,17+,18-,19+,20-,22+,23-,24+/m1/s1

InChI key

QEEBRPGZBVVINN-BMPKRDENSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai Rui LingHu et al.
OncoTargets and therapy, 13, 4767-4778 (2020-06-26)
Bufalin is a component of Chinese traditional medicine, Chansu, which is reported to induce cell death among various kinds of tumors. Apoptosis evasion is a common problem of cancer treatment. The proliferation of U-87 and U-373 treated by bufalin combined
Ze-Yi Zheng et al.
Cancer cell, 37(3), 387-402 (2020-03-07)
We report that neurofibromin, a tumor suppressor and Ras-GAP (GTPase-activating protein), is also an estrogen receptor-α (ER) transcriptional co-repressor through leucine/isoleucine-rich motifs that are functionally independent of GAP activity. GAP activity, in turn, does not affect ER binding. Consequently, neurofibromin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service