Skip to Content
Merck
All Photos(1)

Key Documents

PHL89287

Thymol

phyproof® Reference Substance

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€231.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€231.00

About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907135
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€231.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

vapor pressure

1 mmHg ( 64 °C)

product line

phyproof® Reference Substance

Assay

≥95.0% (GC)

form

crystals

manufacturer/tradename

PhytoLab

bp

232 °C (lit.)

mp

48-51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

InChI key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: ginkgo thymus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service