Skip to Content
Merck
All Photos(1)

Key Documents

PHL83510

Hydroxysafflor yellow A

phyproof® Reference Substance

Synonym(s):

2,4-Di-β-D-glucopyranosyl-3,4,5-trihydroxy-6-[(2E)-3-(4-hydroxyphenyl)-1-oxo-2-propen-1-yl]-2,5-cyclohexadien-1-one, HSYA, Safflomin A

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€379.00

€379.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€379.00

About This Item

Empirical Formula (Hill Notation):
C27H32O16
CAS Number:
Molecular Weight:
612.53
UNSPSC Code:
41116107
NACRES:
NA.24

€379.00


Estimated to ship on08 May 2025


Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

InChI

1S/C27H32O16/c28-7-12-16(32)19(35)21(37)23(42-12)15-18(34)14(11(31)6-3-9-1-4-10(30)5-2-9)24(39)27(41,25(15)40)26-22(38)20(36)17(33)13(8-29)43-26/h1-6,12-13,16-17,19-23,26,28-38,41H,7-8H2/b6-3+,14-11+/t12-,13-,16-,17-,19+,20+,21-,22-,23+,26-,27?/m1/s1

InChI key

IAVUBSCVWHLRGE-UXEKTNMQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maosheng Lee et al.
Diabetes, metabolic syndrome and obesity : targets and therapy, 13, 1097-1107 (2020-04-21)
Type 2 diabetes mellitus (T2DM), a complex metabolic disease, has become a major public health issue around the world. Hydroxysafflor yellow A (HSYA) is the major active chemical ingredient of Carthamus tinctorius L. (safflower), which is widely used in patients with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service