Skip to Content
Merck
All Photos(1)

Key Documents

PHL83363

Carvacrol methyl ether

phyproof® Reference Substance

Synonym(s):

2-Methoxy-p-cymol, 4-Isopropyl-2-methoxytoluene, 5-Isopropyl-2-methylanisole, Methylcarvacrol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€251.00

€251.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€251.00

About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Beilstein:
2412706
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€251.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

liquid

manufacturer/tradename

PhytoLab

density

0.936 g/mL at 20 °C (lit.)

SMILES string

CC1=C(OC)C=C(C(C)C)C=C1

InChI

1S/C11H16O/c1-8(2)10-6-5-9(3)11(7-10)12-4/h5-8H,1-4H3

InChI key

YVLHTQPPMZOCOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: thymus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hatice Büşra Konuk et al.
Folia microbiologica, 65(4), 775-783 (2020-03-21)
Terpenoids, one of the major components of essential oils, are known to exert potent antifungal activity against yeast Saccharomyces cerevisiae. They have been the subject of a considerable number of investigations that uncovered extensive pharmacological properties, including antifungal and antibacterial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service