Skip to Content
Merck
All Photos(1)

Key Documents

PHL83252

Apocynin

phyproof® Reference Substance

Synonym(s):

4′-Hydroxy-3′-methoxyacetophenone, Acetovanillone, Apocynin

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€231.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€231.00

About This Item

Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
Beilstein:
637373
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€231.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

bp

263-265 °C/17 mmHg (lit.)

mp

112-115 °C (lit.)

SMILES string

COc1cc(ccc1O)C(C)=O

InChI

1S/C9H10O3/c1-6(10)7-3-4-8(11)9(5-7)12-2/h3-5,11H,1-2H3

InChI key

DFYRUELUNQRZTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Ku et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 71(3) (2020-10-21)
Fatty acid ethyl esters (FAEEs), non-oxidative metabolites of ethanol, are the main causative agents of severe acute pancreatitis resulting from alcohol abuse. Pancreatic acinar cells exposed to ethanol in combination with the fatty acid palmitoleic acid (EtOH/POA) display increased levels

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service