Skip to Content
Merck
All Photos(1)

Key Documents

PHL82299

Curcumol

phyproof® Reference Substance

Synonym(s):

(−)-Curcumol, (3S,3aS,5S,6R,8aS)-Octahydro-3-methyl-8-methylene-5-(1-methylethyl)-6H-3a,6-epoxyazulen-6-ol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€385.00

€385.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€385.00

About This Item

Empirical Formula (Hill Notation):
C15H24O2
CAS Number:
Molecular Weight:
236.35
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€385.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

SMILES string

CC(C)[C@@H]1C[C@@]23O[C@]1(O)CC(=C)[C@@H]2CC[C@@H]3C

InChI

1S/C15H24O2/c1-9(2)13-8-14-11(4)5-6-12(14)10(3)7-15(13,16)17-14/h9,11-13,16H,3,5-8H2,1-2,4H3/t11-,12-,13-,14-,15+/m0/s1

InChI key

QRMPRVXWPCLVNI-YYFQZIEXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: curcuma

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service