Skip to Content
Merck
All Photos(1)

Key Documents

PHL80322

Trioxsalen

phyproof® Reference Substance

Synonym(s):

2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one, 4,5′,8-Trimethylpsoralen, TMP, Trisoralen

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€231.00

€231.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
Beilstein:
221723
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€231.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

mp

229-231 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cc2cc3C(C)=CC(=O)Oc3c(C)c2o1

InChI

1S/C14H12O3/c1-7-4-12(15)17-14-9(3)13-10(6-11(7)14)5-8(2)16-13/h4-6H,1-3H3

InChI key

FMHHVULEAZTJMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Photochemical crosslinker of DNA that has been used as a probe for nucleic acid structure and function.
Photochemical crosslinker of DNA that has been used as a probe for nucleic acid structure and function. Trioxsalen has also been used to crosslink DNA onto mica surfaces.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Arigò et al.
Journal of chromatography. A, 1649, 462183-462183 (2021-05-27)
In the European Union, cosmetic products are regulated by the Cosmetics Regulation (EC) No. 1223/2009. The Analytical Working Group of the International Fragrance Association (IFRA) suggested that a new sensitive analytical method is needed to determine psoralens in finished cosmetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service