Skip to Content
Merck
All Photos(1)

Key Documents

PHL80267

Plumbagin

phyproof® Reference Substance

Synonym(s):

2-Methyljuglone, 5-Hydroxy-2-methyl-1,4-naphthoquinone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€252.00

€252.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€252.00

About This Item

Empirical Formula (Hill Notation):
C11H8O3
CAS Number:
Molecular Weight:
188.18
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€252.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

Oc1c2c(ccc1)C(=O)C(=CC2=O)C

InChI

1S/C11H8O3/c1-6-5-9(13)10-7(11(6)14)3-2-4-8(10)12/h2-5,12H,1H3

InChI key

VCMMXZQDRFWYSE-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Arati P Vasav et al.
Fitoterapia, 147, 104761-104761 (2020-10-19)
Plumbagin is a pharmacologically active naphthoquinone present in the Plumbago zeylanica L. having important medicinal properties. The root of P. zeylanica is rich and primary tissue of the plumbagin biosynthesis and accumulation. The complete biosynthetic pathway of plumbagin in plant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service