Skip to Content
Merck
All Photos(1)

Key Documents

PHL80142

Maleic acid

phyproof® Reference Substance

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€221.00

€221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€221.00

About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein:
605762
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€221.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

mp

130-135 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service