Skip to Content
Merck
All Photos(1)

Key Documents

PHL80020

Sorbic acid

phyproof® Reference Substance

Synonym(s):

α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€231.00

€231.00


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€231.00

About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098547
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€231.00


Estimated to ship on12 April 2025


Request a Bulk Order

grade

primary reference standard

vapor pressure

0.01 mmHg ( 20 °C)

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

crystals

manufacturer/tradename

PhytoLab

mp

132-135 °C (lit.)

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service