Skip to Content
Merck
All Photos(1)

Key Documents

N12482

Metsulfuron-methyl

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N5O6S
CAS Number:
Molecular Weight:
381.36
Beilstein:
587472
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. N-12482-100MG

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

COC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2

InChI

1S/C14H15N5O6S/c1-8-15-12(18-14(16-8)25-3)17-13(21)19-26(22,23)10-7-5-4-6-9(10)11(20)24-2/h4-7H,1-3H3,(H2,15,16,17,18,19,21)

InChI key

RSMUVYRMZCOLBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Peng et al.
Analytica chimica acta, 674(2), 190-200 (2010-08-04)
This paper reports the preparation of metsulfuron-methyl (MSM) imprinted polymer layer-coated silica nanoparticles toward analysis of trace sulfonylurea herbicides in complicated matrices. To induce the selective occurrence of surface polymerization, the polymerizable double bonds were first grafted at the surface
J D M Belgers et al.
Chemosphere, 85(6), 1017-1025 (2011-08-31)
We tested the effects of the herbicide metsulfuron-methyl on growth of the submerged macrophyte Myriophyllum spicatum under laboratory conditions using different exposure scenarios. The exposures of each scenario were comparable in the concentration × time factor, viz., the same 21-d
B S Ismail et al.
Journal of environmental biology, 33(3), 573-577 (2012-10-04)
Laboratory experiments were conducted to evaluate adsorption, desorption and mobility of metsulfuron-methyl in soils of the oil palm agroecosystem consisting of the Bernam, Selangor, Rengam and Bongor soil series. The lowest adsorption of metsulfuron-methyl occurred in the Bongor soil (0.366
Qiang Tang et al.
Journal of environmental sciences (China), 24(8), 1492-1497 (2012-01-01)
Tribenuron methyl (TBM) is a member of the sulfonylurea herbicide family and is widely used worldwide. In this study, TBM-degrading bacteria were enriched with TBM as potential carbon, nitrogen or sulfur source, and 44 bacterial isolates were obtained. These isolates
Alexandra Pretto et al.
Ecotoxicology and environmental safety, 74(3), 336-341 (2010-11-03)
The effects of commercial formulation containing quinclorac and metsulfuron-methyl herbicides on acetylcholinesterase (AChE), antioxidant profile and metabolic parameters in teleost fish (Leporinus obtusidens) were studied. The fish were exposed during 90 days to commercial formulation containing quinclorac (204 μg L(-1))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service