Skip to Content
Merck
All Photos(1)

Key Documents

KRSU17

Sulfur in Odourless Kerosene

2.0000% (20′000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€140.00

€140.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€140.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€140.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material (UKAS ISO 17025 & ISO 17034 certified)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

concentration

2.0000% (20′000 ppm)

application(s)

petroleum

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
KRSUBLKKRSU20KRSU02
format

matrix material

format

matrix material

format

matrix material

format

matrix material

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum

application(s)

petroleum

application(s)

petroleum

application(s)

petroleum

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

concentration

2.0000% (20′000 ppm)

concentration

-

concentration

5.0000% (50′000 ppm)

concentration

0.0010% (10 ppm)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service