Skip to Content
Merck
All Photos(1)

Key Documents

FAP80868

Aflas in maize

reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€265.00

€265.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€265.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

€265.00


Check Cart for Availability

Request a Bulk Order

grade

reference material

Quality Level

manufacturer/tradename

Fera Science Ltd

application(s)

food and beverages

format

matrix material

storage temp.

−20°C

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Biotoxins / Mycotoxins
Aflatoxin B1, Aflatoxin B2, Aflatoxin G1, Aflatoxin G2, Aflatoxin (total)

Matrix Group: Cereals, Bread, Rice and Maize

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service