Skip to Content
Merck
All Photos(1)

Key Documents

ERMFD103

Titanium dioxide nanorods, suspension in 1-butanol

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€884.00

€884.00


Estimated to ship on28 April 2025


Request a Bulk Order

Select a Size

Change View
1 EA
€884.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€884.00


Estimated to ship on28 April 2025


Request a Bulk Order

grade

certified reference material

Quality Level

Agency

ERM®

manufacturer/tradename

JRC

application(s)

general analytical

Legal Information

ERM is a registered trademark of European Commission

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. do you offer tio2 nano rods? what are the dimensions (diameter and length)? state (powder or suspension)?

    1 answer
    1. This product is a nanorod suspension in 1-butanol. The particles have a median diameter range of 16 nm - 54 nm with a median aspect ratio of 0.296. The specific dimensions (diameter and length) of the nanorods can be found on the certificate found on the European Commission website:
      https://crm.jrc.ec.europa.eu/p/ERM-FD103

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service