Skip to Content
Merck
All Photos(1)

Documents

ERMBF446A

Maize GMO Standard ERM-BF446

certified reference material, 0% (MIR162) Maize for the detection of GMO maize (MIR162) with PCR, ERM®

Synonym(s):

Maize MIR162, GMO Standard ERM-BF446

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

food and beverages

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please click on the below product number:
ERMBF446A

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service