D2622LMPSB
Polysulfide in light mineral oil, ASTM D2622
Calibration Set B
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Quality Level
form
liquid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
Looking for similar products? Visit Product Comparison Guide
Components
Kit contains: LMPS110, LMPS120, LMPS140, LMPS150; 100 mL ea.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service