Skip to Content
Merck
All Photos(1)

Key Documents

CRM96142

Supelco

Bis[(2,3-epoxypropoxy)phenyl] methane

certified reference material, TraceCERT®, mixture of isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€139.00

€139.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€139.00

About This Item

Empirical Formula (Hill Notation):
C19H20O4
CAS Number:
Molecular Weight:
312.36
UNSPSC Code:
41116107
NACRES:
NA.24

€139.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CCC(C)CC.CC.CC.CC(CC)CC.C1(CC2=CC=CC=C2)=CC=CC=C1.O.O.O.O

InChI

1S/C19H20O4/c1-3-7-18(22-12-16-10-20-16)14(5-1)9-15-6-2-4-8-19(15)23-13-17-11-21-17/h1-8,16-17H,9-13H2

InChI key

FIJSKXFJFGTBRV-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Linkage

Ion Mobility: TWCCSN2 value of 177.5 Å2 [M+H]+ and 168.2 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N CRM96142 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Analysis Note

This product contains ortho-ortho, ortho-para and para-para isomers

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service