Skip to Content
Merck
All Photos(1)

Key Documents

CRM80839

Supelco

Tritosulfurone Metabolite M635H004

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Amino-4-methoxy-6-(trifluoromethyl)-1,3,5-triazine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€106.00

€106.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€106.00

About This Item

Empirical Formula (Hill Notation):
C5H5F3N4O
CAS Number:
Molecular Weight:
194.11
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€106.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)c1nc(nc(n1)OC)N

InChI

1S/C5H5F3N4O/c1-13-4-11-2(5(6,7)8)10-3(9)12-4/h1H3,(H2,9,10,11,12)

InChI key

DTVMLDILWYLGLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Tritosulfurone Metabolite M635H004 is formed from the use of a new active substance tritosulfuron, which belongs to the chemical class of sulfonylureas. Tritosulfuron― a systemic herbicide, is an acetolactate synthase (ALS) inhibitor. It is used for the post-emergence control of a variety of dicotyledonous weeds in cereals and maize.
As per Regulation (EC) No. 1107/2009, repealing directive 91/414/EEC, tritosulfuron is approved for use in the European Union as a plant protection agent.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloride promoted oxidation of tritosulfuron by peroxymonosulfate
Rivas FJ and Solis RR
Chemical Engineering Journal, 349, 728-736 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service